Sample containers with screw cap, Bijou, WHEATON®
7 ml, Bijou container, plain label, sterile, Clear, PS/PE
PS clear, with PE screw cap.
- Manufactured from FDA approved virgin PS
- Supplied sterile and for in vitro use only
- PS offers good thermal and chemical resistance
- All materials are non-cytotoxic
Straight-walled vessels ideal for sample collection, storage and transport. Supplied with leak proof, flow seal closure. Available pre-filled with boric acid for urine samples.
: CE marked in accordance with European Directive 98/79/EC
All containers are leak tested to British Standard BS5213:1975(1989)
: Packed in inner bags of 50.
: *The product(s) on this page are outside the scope of the EU Directive 2019/904
- Type:Bijou container, plain label, sterile
- Capacity:7 ml
- Colour:Clear
- Material:PS/PE