ContactUsLinkComponent
ARROW NUKLEINSÄURE-EXTRAKTIONS-AUTOMAT (CE, IVD) Specifications: Number of samples: 1-12 Throughput for 1-12 samples: >30 min, depending on protocol Dimensions (WxDxH): 46x47x51 cm Weight: 17 Kg Electrical: 100-240 VAC, 50-60 Hz Interfaces: USB 1.1, 1 * 1 ST
  IBSA8.31.02
ARROW NUKLEINSÄURE-EXTRAKTIONS-AUTOMAT (CE, IVD) Specifications: Number of samples: 1-12 Throughput for 1-12 samples: >30 min, depending on protocol Dimensions (WxDxH): 46x47x51 cm Weight: 17 Kg Electrical: 100-240 VAC, 50-60 Hz Interfaces: USB 1.1, 1 * 1 ST
  IBSA8.31.02