Orders LinkContactUsLinkComponent
BOD measurement
BOD measurement
BOD Sensor only
Catalog # VELPF102B0133
Supplier:   VELP SCIENTIFIC
BOD measurement
Catalog # VELPF102B0133
Supplier:  VELP SCIENTIFIC
BOD measurement
BOD measurement
BOD Sensor only
Price
268,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Measurement system without mercury for determination of the amount of dissolved oxygen consumed by aerobic biological microorganisms in water. The sensor uses manometric technology and micrprocessor control to provide regular data measurements over defined periods.
  • Very simple to use and mercury-free
  • Five values every 24 hours can be memorised or viewed instantaneously on the numerical display
  • 1, 6 and 10 sample bottle versions available
  • Measurement ranges: 90/250/600/999 ppm BOD
Temperature control is usually required for reproducible results - please see entries for thermostatic cabinets and incubators which are supplied separately.
Temperature control is usually required for reproducible results - please see entries for thermostatic cabinets and incubators which are supplied separately.
Delivery information: The BOD systems includes sensors, dark glass sample bottles, alkali holder for absorption of CO2, stirrer platform (not for single bottle vesrion) and stirrer bars.
Documents
Specifications
  • Description:
    BOD Sensor only
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Dark glass bottle 500 ml for BOD systems
    Dark glass bottle 500 ml for BOD systems
    705-0781
    Log in for availability
    1 * 1 items - 13,20 €
Product Information
  • Catalog #
    Availability
    Price
  • Control test tablets
    Control test tablets
    705-0782
    Log in for availability
    1 * 1 items - 79,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Potassium hydroxide trap for BOD sensor
    Potassium hydroxide trap for BOD sensor
    705-0845
    Log in for availability
    1 * 1 items - 9,60 €