Orders LinkContactUsLinkComponent
Flasks, Erlenmeyer, with standard ground joint
Flasks, Erlenmeyer, with standard ground joint
150 ml, 24/29 NS
Catalog # SMIT8SFE150/B24
Supplier:   SMITH SCIENTIFIC
Flasks, Erlenmeyer, with standard ground joint
Catalog # SMIT8SFE150/B24
Supplier:  SMITH SCIENTIFIC
Flasks, Erlenmeyer, with standard ground joint
150 ml, 24/29 NS
Price
1 230,00 €
Log in for availability
1 * 6 items
Product Details & Documents
Borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Specifications
  • Socket size:
    24/29 NS
  • Capacity:
    150 ml
  • Seq:
    04