Orders LinkContactUsLinkComponent
Flasks, Erlenmeyer, with standard ground joint
Flasks, Erlenmeyer, with standard ground joint
500 ml, 19/26 NS
Catalog # SMIT8SFE500/B19
Supplier:   SMITH SCIENTIFIC
Flasks, Erlenmeyer, with standard ground joint
Catalog # SMIT8SFE500/B19
Supplier:  SMITH SCIENTIFIC
Flasks, Erlenmeyer, with standard ground joint
500 ml, 19/26 NS
Price
1 510,00 €
Log in for availability
1 * 6 items
Product Details & Documents
Borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Specifications
  • Socket size:
    19/26 NS
  • Capacity:
    500 ml
  • Seq:
    14