Ethanolamine 99+% ACS
4
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99+% CAS No.: 141-43-5 Molecular Formula: HOCH2CH2NH2 Formula Weight: 61.08 Flash Point: 93°(199°F) Physical Form: Liquid Density: 1.012 Refractive Index: 1.4540 Sensitivity: Air Sensitive and Hygroscopic Odour: Ammonia-like Melting Point: 10° Boiling Point: 169-170° Beilstein Reference: 505944 Merck Reference: 14,3727 UN No.: 2491 MDL No.: MFCD00008183
Synonyms: 2-Aminoethanol, Monoethanolamine
- CAS Number:141-43-5
- Formula:NH₂CH₂CH₂OH
- MDL Number:MFCD00008183
- Molecular Weight:61.08 g/mol
- Merck Index:14,3727
- Boiling Pt:169…170 °C
- Melting Pt:10 °C
- Density:1.012
- Flash Pt:93 °C (199 °F)
- Storage Temperature:Ambient
- EINECS:205-483-3
- UN:2491
- ADR:8,III
Documents
Specifications
Specifications
- Pk:4



