Kaempferol
Glass bottle for solids, 25 mg
Catalog # CAYM11852-25
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
Synonyms: 3,5,7-Trihydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one
- CAS Number:520-18-3
- Formula:C₁₅H₁₀O₆
- MDL Number:MFCD00016938
- Molecular Weight:286.24 g/mol
- EINECS:208-287-6
Documents
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 mg



