Orders LinkContactUsLinkComponent
QuEChERS extraction salt packets Q-sep™
QuEChERS extraction salt packets Q-sep™
Q-sep QuEChERS extraction Q110 kit (European EN 15662 method) includes 4 g magnesium sulphate, 1 g sodium chloride, 1 g trisodium citrate dihydrate, 0,5 g disodium hydrogen citrate sesquihydrate and 50 ml centrifuge tube
Catalog # REST26235
QuEChERS extraction salt packets Q-sep™
Catalog # REST26235
Supplier:  
QuEChERS extraction salt packets Q-sep™
QuEChERS extraction salt packets Q-sep™
Q-sep QuEChERS extraction Q110 kit (European EN 15662 method) includes 4 g magnesium sulphate, 1 g sodium chloride, 1 g trisodium citrate dihydrate, 0,5 g disodium hydrogen citrate sesquihydrate and 50 ml centrifuge tube
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Q-sep QuEChERS extraction EN Method 4g MgSO4 1g NaCl 1g TSCD 0.5g DHS packets and tubes 1 * 50 items Catalog #REST25850
Product REST25850 is similar to REST26235 but manufacturer and specifications may vary.
Product Details & Documents
Q-sep™ QuEChERS provides fast, simple sample preparation for multi residue pesticide analysis.
  • Preweighed, ultra-pure sorbents
  • Convenient and easy to use
  • Salt packets eliminate the need for a second empty tube to transfer salts
Support original unbuffered, AOAC (2007.01) and European (EN 15662) QuEChERS methods.
Specifications
  • Description:
    Q-sep QuEChERS extraction Q110 kit (European EN 15662 method) includes 4 g magnesium sulphate, 1 g sodium chloride, 1 g trisodium citrate dihydrate, 0,5 g disodium hydrogen citrate sesquihydrate and 50 ml centrifuge tube