Robotic tips for Eppendorf epMotion® automated pipetting system
1000 µl, +, -, 10 racks, 96 each
736-0762
Robotic tips for Eppendorf epMotion® automated pipetting system
736-0762
These automation tips for the epMotion automated pipetting system are available in non sterile, sterile, and sterile filtered versions.
- Each tip is lot-tested during production using a 15-point QC program on an epMotion workstation
- Tips have a low percentage Coefficient of Variation (% CV)
- Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
- Packed:10 racks, 96 each
- Sterile:+
- Volume:1000 µl
- Filter:-
- Cat. No.:736-0762
- Environmental certification:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Environmentally Preferable:Low Manufacturing Impact