Orders LinkContactUsLinkComponent
Robotic tips for Eppendorf epMotion® automated pipetting system
Robotic tips for Eppendorf epMotion® automated pipetting system
50 µl, +, -, 10 racks, 96 each
Catalog # 736-0755
   Low Manufacturing Impact
Robotic tips for Eppendorf epMotion® automated pipetting system
Catalog # 736-0755
   Low Manufacturing Impact
Robotic tips for Eppendorf epMotion® automated pipetting system
Robotic tips for Eppendorf epMotion® automated pipetting system
50 µl, +, -, 10 racks, 96 each
Price
148,00 €
Log in for availability
1 * 960 items
Product Details & Documents
These automation tips for the epMotion automated pipetting system are available in non sterile, sterile, and sterile filtered versions.
  • Each tip is lot-tested during production using a 15-point QC program on an epMotion workstation
  • Tips have a low percentage Coefficient of Variation (% CV)
  • Sterile tips are certified free of RNase, DNase, DNA, endotoxin, bioburden and pyrogen
Specifications
  • Packed:
    10 racks, 96 each
  • Sterile:
    +
  • Volume:
    50 µl
  • Filter:
    -
  • Cat. No.:
    736-0755
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Environmentally Preferable:
    Low Manufacturing Impact