Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Poroshell 120 SB-Aq, 2.1 x 150 mm, 2.7UMLC column 1 * 1 items
Catalog # HEWL683775-914
Supplier:
Agilent
Expand image
Poroshell 120 SB-Aq, 2.1 x 150 mm, 2.7UMLC column 1 * 1 items
Catalog # HEWL683775-914
Supplier:
Agilent