Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Poroshell 120 SB-Aq, 3.0 x 100 mm, 2.7UMLC column 1 * 1 items
Catalog # HEWL685975-314
Supplier:
Agilent
Expand image
Poroshell 120 SB-Aq, 3.0 x 100 mm, 2.7UMLC column 1 * 1 items
Catalog # HEWL685975-314
Supplier:
Agilent