Order Entry
Export
Orders LinkContactUsLinkComponent
Hydrometers for milk

Hydrometers for milk

Hydrometer for milk, 1,025 − 1,035 g/ml, 0,0005 g/ml
Catalog # ALLA2220FC025/20QP
Supplier:   Alla France
Hydrometers for milk
Catalog # ALLA2220FC025/20QP
Supplier:  Alla France
Hydrometers for milk
Hydrometers for milk

Hydrometer for milk, 1,025 − 1,035 g/ml, 0,0005 g/ml

Price
89,70 €
Log in for availability
1 * 1 items
Product Details & Documents

Certifications: BS 734

Specifications
  • Description:
    Hydrometer for milk
  • Seq:
    001
  • Division:
    0,0005 g/ml
  • Range:
    1,025 − 1,035 g/ml