ContactUsLinkComponent
NanoLC-Ultra 1D system. Gradient pump for 50-500 nl/min. Max. pressure 10,000 psi. Temperature controlled column compartment. Includ es NanoLC start-up kit (p/n 801-00043) and Eksigent control software. Installation and training included does not i 1 * 1 items
Catalog # 903-0620
Supplier:  EKSIGENT
NanoLC-Ultra 1D system. Gradient pump for 50-500 nl/min. Max. pressure 10,000 psi. Temperature controlled column compartment. Includ es NanoLC start-up kit (p/n 801-00043) and Eksigent control software. Installation and training included does not i 1 * 1 items
Catalog # 903-0620
Supplier:  EKSIGENT