Formamide ≥99.5% ACS
4
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99.5+% CAS No.: 75-12-7 Molecular Formula: CH3NO Formula Weight: 45.04 Flash Point: 154°(310°F) Physical Form: Liquid Density: 1.134 Refractive Index: 1.4475 Sensitivity: Hygroscopic Appearance: Colourless Odour: Ammonia-like Melting Point: 2° Boiling Point: 210° Beilstein Reference: 505995 Merck Reference: 14,4237 MDL No.: MFCD00007941
Synonyms: Carbamaldehyde
- CAS Number:75-12-7
- Formula:HCONH₂
- MDL Number:MFCD00007941
- Molecular Weight:45.04 g/mol
- Merck Index:14,4237
- Boiling Pt:210 °C
- Melting Pt:2 °C
- Density:1.134
- Flash Pt:154 °C (310 °F)
- Storage Temperature:Ambient
- EINECS:200-842-0
Documents
Specifications
Specifications
- Pk:4



