Orders LinkContactUsLinkComponent
Sample bottles for ASE® 100/300 systems
Sample bottles for ASE® 100/300 systems
250 ml, Clear
Catalog # REST26191
Supplier:   Restek
Sample bottles for ASE® 100/300 systems
Catalog # REST26191
Supplier:  Restek
Sample bottles for ASE® 100/300 systems
Sample bottles for ASE® 100/300 systems
250 ml, Clear
Price
45,00 €
Log in for availability
1 * 12 items
Product Details & Documents
Sample collection bottles are made of glass. Caps and PTFE-lined septa included. Suitable for ASE 100/300 systems.
Specifications
  • Capacity:
    250 ml
  • Colour:
    Clear