Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
GHS HAZARDOUS MATERIALS SIGNS No. 5, 13x 1 * 10 items
Catalog # LBIN9910-GHS05
Supplier:
LAB IN
Expand image
GHS HAZARDOUS MATERIALS SIGNS No. 5, 13x 1 * 10 items
Catalog # LBIN9910-GHS05
Supplier:
LAB IN