VWR® Buffer Solutions, Secondary Standards
12,454, ±0,050, 25 °C, Plastic bottle, 500 mL
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
- Accurate to ±0,010 pH units (except buffer 12,454 with accuracy ±0,050)
- Solution standardised at 25 °C
- Fully traceable for optimum pH calibration (traceable to NIST, DIN 19266 and IUPAC)
- Temperature dependence data is printed on the labels as are batch number and expiry date
Specifications
Specifications
- Pack type:Plastic bottle
- Pk:500 mL
- Reference temperature:25 °C
- pH accuracy:±0,050
- pH value:12,454



