Order Entry
Export
Orders LinkContactUsLinkComponent
VWR® Buffer Solutions, Secondary Standards

VWR® Buffer Solutions, Secondary Standards

7,413, ±0,010, 25 °C, Plastic bottle, 500 mL
Catalog # 84584.260
Supplier:   VWR Chemicals
VWR® Buffer Solutions, Secondary Standards
Catalog # 84584.260
Supplier:  VWR Chemicals
CAS Number:  
VWR® Buffer Solutions, Secondary Standards
VWR® Buffer Solutions, Secondary Standards

7,413, ±0,010, 25 °C, Plastic bottle, 500 mL

CAS Number:  
Price
57,10 €
Log in for availability
1 * 500 mL
Product Details & Documents
  • Accurate to ±0,010 pH units (except buffer 12,454 with accuracy ±0,050)
  • Solution standardised at 25 °C
  • Fully traceable for optimum pH calibration (traceable to NIST, DIN 19266 and IUPAC)
  • Temperature dependence data is printed on the labels as are batch number and expiry date
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Reference temperature:
    25 °C
  • pH accuracy:
    ±0,010
  • pH value:
    7,413

Specifications