Orders LinkContactUsLinkComponent
Film applicators, multi-functional
Film applicators, multi-functional
Multi-functional film applicator, 4 sided, gaps at 15, 30, 60, 90 µm, width 60 mm
Catalog # SIMEVF2168
Supplier:   Industrial Physics
Film applicators, multi-functional
Catalog # SIMEVF2168
Supplier:  Industrial Physics
Film applicators, multi-functional
Film applicators, multi-functional
Multi-functional film applicator, 4 sided, gaps at 15, 30, 60, 90 µm, width 60 mm
Price
555,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Multi-functional film applicator with 4 application sides for applying paint films of 4 different predefined thicknesses. The applicators are available in film widths of 60, 80 mm and one side of the applicator is supplied with a guidance support for straight application. Made from high grade stainless steel, the multi-functional film applicator will not be affected by acid or base elements.
Documents
Specifications
  • Description:
    Multi-functional film applicator, 4 sided, gaps at 15, 30, 60, 90 µm, width 60 mm
  • Seq:
    02