Immersion flow cups, ISO 2431
Viscosity immersion flow cup, titanium anodized aluminium, 8 mm
Catalog # 649-0010
Supplier: Industrial Physics
Jump to:
Product Details & Documents
Titanium anodized aluminum or stainless steel cups designed with fixed stainless steel nozzle and handle, for measuring coatings and other fluids.
Certifications: ISO 2431
Documents
TQC Company Video
Videos
Specifications
- Description:Viscosity immersion flow cup, titanium anodized aluminium
- Seq:05
- Cup orifice:8 mm
Accessories
Product Information
- Catalog #AvailabilityPrice
- Temperature control jacket with tripod for ISO and AFNOR cupsTemperature control jacket with tripod for ISO and AFNOR cupsSIMEVF2068Log in for availability1 * 1 items - 627,00 €
Product Information
- Catalog #AvailabilityPrice
- Digital stopwatch, 0,01 s
Product Information
- Catalog #AvailabilityPrice
- Viscosity convertor of flow time to viscosityViscosity convertor of flow time to viscosity769-1151Log in for availability1 * 1 items - 87,50 €



