Orders LinkContactUsLinkComponent
Lab Markers, several colours
Lab Markers, several colours
Lab marker, fine tip, Black
Catalog # PRCI1400-20-PDC
Supplier:   PRECISION DYNAMICS
Lab Markers, several colours
Catalog # PRCI1400-20-PDC
Supplier:  PRECISION DYNAMICS
Lab Markers, several colours
Lab marker, fine tip, Black
Price
29,70 €
Log in for availability
1 * 10 items
Product Details & Documents
Versatile, permanent markings write on plastic, metal, and porcelain labware.
  • Several colours
  • Unique ink has the ability to write on labware even when the surface is damp
Specifications
  • Colour:
    Black
  • Description:
    Lab marker, fine tip