Lab Markers, several colours
Lab marker, fine tip, Blue
Catalog # PRCI1400-13-PDC
Supplier: PRECISION DYNAMICS
Jump to:
Product Details & Documents
Versatile, permanent markings write on plastic, metal, and porcelain labware.
- Several colours
- Unique ink has the ability to write on labware even when the surface is damp
Specifications
- Colour:Blue
- Description:Lab marker, fine tip
- Seq:01