Orders LinkContactUsLinkComponent
Filtering apparatus, DURAN®
Complete with 1000 ml filtration bottle, 250 ml filter head, PTFE insert and clamp
Catalog # 516-0431
Supplier:   DWK Life Sciences
Filtering apparatus, DURAN®
Catalog # 516-0431
Supplier:  DWK Life Sciences
Filtering apparatus, DURAN®
Filtering apparatus, DURAN®
Complete with 1000 ml filtration bottle, 250 ml filter head, PTFE insert and clamp
Price
685,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Filtration apparatus with unit made from clear DURAN® borosilicate glass 3.3, PTFE insert and anodised aluminium clamp.
  • Scaled funnel
  • All components are available as spare parts
Typical applications: Coarse and ultrafine filtration, filtration of HPLC media, residue analysis. Filter paper, membrane filters (47 mm) or glass filters can be used for filtration.
Specifications
  • Description:
    Complete with 1000 ml filtration bottle, 250 ml filter head, PTFE insert and clamp
  • Seq:
    01
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Clamp, anodised aluminium
    Clamp, anodised aluminium
    SCOT290763609
    Log in for availability
    1 * 1 items - 141,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Funnel with 500 ml scale
    Funnel with 500 ml scale
    SCOT247224401
    Log in for availability
    1 * 1 items - 195,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Filtering flask, 2000 ml, NS 45/40
    Filtering flask, 2000 ml, NS 45/40
    SCOT242026306
    Log in for availability
    1 * 1 items - 208,00 €
Product Information
  • Catalog #
    Material
    Ext.Ø
    Availability
    Price
  • Hose connection, straight, with silicone seal (VMQ)
    Hose connection, straight, with silicone seal (VMQ)
    201-2101
    PP
    8.6 mm
    Log in for availability
    1 * 1 items - 1,70 €
Product Information
  • Catalog #
    Thread
    Colour
    Availability
    Price
  • Screw cap with aperture, height: 28 mm, bore: 34 mm
    Screw cap with aperture, height: 28 mm, bore: 34 mm
    201-1921
    GL 14
    Red
    Log in for availability
    1 * 1 items - 2,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Adapter, PTFE, NS 45/40 - GL45
    Adapter, PTFE, NS 45/40 - GL45
    SCOT294001207
    Log in for availability
    1 * 1 items - 150,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Head, NS 45/40
    Head, NS 45/40
    SCOT243173203
    Log in for availability
    1 * 1 items - 174,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Funnel with 250 ml scale
    Funnel with 250 ml scale
    SCOT247223602
    Log in for availability
    1 * 1 items - 172,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Filtering flask, 1000 ml, NS 45/40
    Filtering flask, 1000 ml, NS 45/40
    SCOT242025404
    Log in for availability
    1 * 1 items - 85,90 €
Product Information
  • Catalog #
    Availability
    Price
  • Adapter disc, PTFE
    Adapter disc, PTFE
    SCOT294002803
    Log in for availability
    1 * 1 items - 115,00 €