Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
PH METER FEP20-ATC KIT + PRINTER RS-P25 1 * 1 items
Catalog # 662-1417
Supplier:
Mettler - Toledo
Expand image
PH METER FEP20-ATC KIT + PRINTER RS-P25 1 * 1 items
Catalog # 662-1417
Supplier:
Mettler - Toledo