Order Entry
Export
Orders LinkContactUsLinkComponent
 

Milk, powder

Glass bottle for solids, 1 kg
Catalog # ROTHT145.3
Supplier:   Roth Carl
Milk, powder
Catalog # ROTHT145.3
Supplier:  Roth Carl
CAS Number:  999999-99-4
Milk, powder

Glass bottle for solids, 1 kg

CAS Number:  999999-99-4
Price
45,60 €
Log in for availability
1 * 1 kg
Product Details & Documents
  • CAS Number:999999-99-4
  • EINECS:310-127-6

 

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications