Horizontal laminar flow, Purifier®
Horizontal laminar flow cabinet, nominal width of 6 feet, with interior depth of 530 mm, UV light, EU-plug
LBCP3873028
: LABCONCO
It offers a choice to protect materials that do not generate harmful aerosols or vapors. Its been designed with 99,99% efficient HEPA filter, the air from this minimizes cross contamination in the work area and provides a particulate free work environment. It has wide range of applications including plant tissue culture research, media preparation, electronic part inspection, syringe filling, medical device assembly and non hazardous parenteral drug formulation. These benches should never be used in conjunction with biohazardous material, toxins or radionuclides.
- Differential pressure gauge
- Fluorescent lighting
- Glacier white powder coated steel exterior
- Available in various width sizes and has a differential pressure gauge that displays pressure across the HEPA filter
Construction: Work surface made of stainless steel
: Air quality Class 5 - ISO 14644-1 (Class 100).
- Description:Horizontal laminar flow cabinet, nominal width of 6 feet, with interior depth of 530 mm, UV light, EU-plug
- Seq:22
- Cat. No.:LBCP3873028
- Filters:HEPA filter: 99,99% efficient
- Lighting:Fluorescent lighting
- Model:Purifier®
- Electrical supply:230 V, 50/60 Hz
- Ext. W×D×H:1905×864×1229 mm
- Weight:293 kg
Services
- Description
- SERW1211.7000.01Installation Qualification and Operation Qualification of LAF at customer site. Qualification Protocols not included. Travel fees included (cost of air travel excluded).1 * 1 Each - 0,00 €
- SERW1211.1101.01Internal check and operational testing (EN 14644) of LAF at customer site. Service report included. Spare parts and travel fees not included1 * 1 Each - 679,00 €
- SERW1211.0100.01Installation of LAF at customer site. The device must be at the place of use. Functionality test and brief user introduction to functions. Travel fees included (cost of air travel excluded).1 * 1 Each - 679,00 €
- SERW1211.0300.01Installation and operational testing (EN 14644) of LAF at customer site. The device must be at the place of use. Spare parts not included. Travel fees included (cost of air travel excluded).1 * 1 Each - 679,00 €
Accessories
- DescriptionW×D×H
- LBCP3746701Telescoping base stands with fixed feet, nominal width of 76,2 mm, 914×737 mm, 43 kg— mm1 * 1 items - 2 150,00 €
- DescriptionW×D×H
- LBCP3746700Telescoping base stands with fixed feet, nominal width of 50,8 mm, 610×737 mm, 36 kg— mm1 * 1 items - 2 040,00 €
- DescriptionW×D×H
- 135-2005Telescoping base stands with fixed feet, nominal width of 101,6 mm, 1194×737 mm, 55 kg— mm1 * 1 items



