Order Entry
Export
Orders LinkContactUsLinkComponent
Horizontal laminar flow, Purifier®

Horizontal laminar flow, Purifier®

Horizontal laminar flow cabinet, nominal width of 4 feet, with interior depth of 530 mm, UV light, UK-plug
Catalog # LBCP3888425
Horizontal laminar flow, Purifier®
Catalog # LBCP3888425
Supplier:  
Horizontal laminar flow, Purifier®
Horizontal laminar flow, Purifier®

Horizontal laminar flow cabinet, nominal width of 4 feet, with interior depth of 530 mm, UV light, UK-plug

Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Bench, clean horizontal, purifier, nominal width 121.9cm, interior depth 53cm, electrical requirement 230V, 50/60hz, 3amps, light fl uorescent/UV, plug schuko. 1 * 1 items Catalog #LBCP3888428

Product LBCP3888428 is a replacement for LBCP3888425 and has the same packaging and specifications, but may be a different quantity.

Product Details & Documents

It offers a choice to protect materials that do not generate harmful aerosols or vapors. Its been designed with 99,99% efficient HEPA filter, the air from this minimizes cross contamination in the work area and provides a particulate free work environment. It has wide range of applications including plant tissue culture research, media preparation, electronic part inspection, syringe filling, medical device assembly and non hazardous parenteral drug formulation. These benches should never be used in conjunction with biohazardous material, toxins or radionuclides.

  • Differential pressure gauge
  • Fluorescent lighting
  • Glacier white powder coated steel exterior
  • Available in various width sizes and has a differential pressure gauge that displays pressure across the HEPA filter

Construction: Work surface made of stainless steel

Certifications: Air quality Class 5 - ISO 14644-1 (Class 100).

Specifications
  • Description:
    Horizontal laminar flow cabinet, nominal width of 4 feet, with interior depth of 530 mm, UV light, UK-plug
  • Seq:
    15
  • Cat. No.:
    LBCP3888425
  • Filters:
    HEPA filter: 99,99% efficient
  • Lighting:
    Fluorescent lighting
  • Model:
    Purifier®
  • Electrical supply:
    230 V, 50/60 Hz
  • Ext. W×D×H:
    1295×864×1229 mm
  • Weight:
    206 kg
Services

Product Information

  • Catalog #
    Description
    Availability
    Price
  • SERW1211.7000.01
    Installation Qualification and Operation Qualification of LAF at customer site. Qualification Protocols not included. Travel fees included (cost of air travel excluded).
    Log in for availability
    1 * 1 Each - 0,00 €

  • SERW1211.1101.01
    Internal check and operational testing (EN 14644) of LAF at customer site. Service report included. Spare parts and travel fees not included
    Log in for availability
    1 * 1 Each - 679,00 €

  • SERW1211.0100.01
    Installation of LAF at customer site. The device must be at the place of use. Functionality test and brief user introduction to functions. Travel fees included (cost of air travel excluded).
    Log in for availability
    1 * 1 Each - 679,00 €

  • SERW1211.0300.01
    Installation and operational testing (EN 14644) of LAF at customer site. The device must be at the place of use. Spare parts not included. Travel fees included (cost of air travel excluded).
    Log in for availability
    1 * 1 Each - 679,00 €

Accessories

Product Information

  • Catalog #
    W×D×H
    Availability
    Price
  • Telescoping base stands with fixed feet, nominal width of 76,2 mm, 914×737 mm, 43 kg
    Telescoping base stands with fixed feet, nominal width of 76,2 mm, 914×737 mm, 43 kg
    LBCP3746701
    — mm
    Log in for availability
    1 * 1 items - 2 150,00 €

Product Information

  • Catalog #
    W×D×H
    Availability
    Price
  • Telescoping base stands with fixed feet, nominal width of 50,8 mm, 610×737 mm, 36 kg
    Telescoping base stands with fixed feet, nominal width of 50,8 mm, 610×737 mm, 36 kg
    LBCP3746700
    — mm
    Log in for availability
    1 * 1 items - 2 040,00 €

Product Information

  • Catalog #
    W×D×H
    Availability
    Price
  • Telescoping base stands with fixed feet, nominal width of 101,6 mm, 1194×737 mm, 55 kg
    Telescoping base stands with fixed feet, nominal width of 101,6 mm, 1194×737 mm, 55 kg
    135-2005
    — mm
    Log in for availability
    1 * 1 items