Order Entry
Export
Orders LinkContactUsLinkComponent
 

Filling and venting closures, Nalgene®

53B, Nalgene bottles and carboys with 53B closures, 6,3 mm
  216-8911
   Low Manufacturing Impact
Filling and venting closures, Nalgene®
  216-8911
   Low Manufacturing Impact
Filling and venting closures, Nalgene®
Filling and venting closures, Nalgene®

53B, Nalgene bottles and carboys with 53B closures, 6,3 mm

 
45,80 €
   
1 * 1 items
 

PP, white with TPE seals and TPE caps.

  • Fit all large Nalgene® carboys or bottles with 53 mm (53 B) or 83 mm (83 B) screw caps
  • Choice of 6,3 mm or 12,7 mm tube for the 83 mm cap (83 B)
  • Autoclavable

Screw closures for the sterile liquid transfer of nutrient media, biological reagents, pure water and chemicals. For applications requiring the use of bioreactors and fermenters.

 : Pack includes the cap, Nalgene® platinum-treated silicone tubing for aspirating and dispensing and a splash guard for filling.

 
  • Cap Size:
    53B
  • For:
    Nalgene bottles and carboys with 53B closures
  • Tubing int.Ø:
    6,3 mm
  • Seq:
    01
  • Cat. No.:
    216-8911
  • Environmental certification:
    NAR Certificate of Retirement
  • Environmentally Preferable:
    Low Manufacturing Impact