Phanerogames prepared slides
Epidermis of Allium cepa (onion), w.m. showing simple plant cells with cell walls, nuclei and cytoplasm
Catalog # LIEDAS111C
Supplier: LIEDER JOHANNES
Jump to:
Product Details & Documents
These seagrass preparations are available sets of five.
Specifications
- Description:Epidermis of Allium cepa (onion), w.m. showing simple plant cells with cell walls, nuclei and cytoplasm
- Seq:02