Phanerogames prepared slides
Tuber of Solanum (potato) t.s. shows cork and starch grains
Catalog # LIEDAS371C
Supplier: LIEDER JOHANNES
Jump to:
Product Details & Documents
These seagrass preparations are available sets of five.
Specifications
- Description:Tuber of Solanum (potato) t.s. shows cork and starch grains
- Seq:07