ContactUsLinkComponent
CTC Analytics PAL DLW-HTC-xt autosampler with DLW-2 wash station configured for use with the ekspert microLC 200 pump. Includes six- plate (three drawer) cooled sample hotel. Installation and training included if ordered as part of a system. 1 * 1 items
Catalog # 903-0606
Supplier:  EKSIGENT
undefined
CTC Analytics PAL DLW-HTC-xt autosampler with DLW-2 wash station configured for use with the ekspert microLC 200 pump. Includes six- plate (three drawer) cooled sample hotel. Installation and training included if ordered as part of a system. 1 * 1 items
Catalog # 903-0606
Supplier:  EKSIGENT