ContactUsLinkComponent
ekspert microLC 200 binary gradient pump configured for 5-50 uL/min flow rate. Includes remotely mounted column oven, electrically-a ctuated low dispersion 6-port injection valve, and mounting brackets for CTC six plate (three drawer) cooled sample h 1 * 1 items
Catalog # 903-0605
Supplier:  EKSIGENT
undefined
ekspert microLC 200 binary gradient pump configured for 5-50 uL/min flow rate. Includes remotely mounted column oven, electrically-a ctuated low dispersion 6-port injection valve, and mounting brackets for CTC six plate (three drawer) cooled sample h 1 * 1 items
Catalog # 903-0605
Supplier:  EKSIGENT