Multi-channel pipettes, variable volume, Axygen® Axypet™
5 - 50, ±4,0 - ±1,6 %, <2,5 - <0,6 %, TR-222 µl
Catalog # AXYGAP-8-50
Supplier: Corning
Jump to:
Product Details & Documents
These pipettes combine the highest levels of accuracy and precision with design innovations intended to make the pipetting experience more comfortable over prolonged periods.
- Digital volume display with easy to read numbers
- Advanced, lightweight materials (body of the pipette and piston assemblies)
- Each channel has an individual precision piston assembly to ensure accuracy and reproducibility from one pipetting series to the next, as well as between channels
- Micrometer is continuously adjustable for selection of whole or fractional volumes
The plunger and ejector buttons are ergonomically located and operate smoothly to allow comfortable pipetting by both right and left handed users. For comfortable pipetting in any direction, the tip manifold rotates 360°. The innovative ejector bar is curved, allowing the tips to be pushed off in steps, therefore reducing the amount of force required for ejection.
Specifications
- Volume:5 - 50
- Seq:02
- Accuracy:±4,0 - ±1,6 %
- Imprecision:<2,5 - <0,6 %
- Recommended tips:TR-222 µl
Accessories
Product Information
- Catalog #AvailabilityPrice
- Carousel stand, holds 6 single channel pipettes (not compatible with Axygen Axypet Pro pipettors)Carousel stand, holds 6 single channel pipettes (not compatible with Axygen Axypet Pro pipettors)AXYGAP-CSDLog in for availability1 * 1 items - 169,00 €
Product Information
- Catalog #AvailabilityPrice
- Universal linear stand, transparent, for four pipettors (four single-channel or max. two 8-channel or 12-channel pipettors)Universal linear stand, transparent, for four pipettors (four single-channel or max. two 8-channel or 12-channel pipettors)AXYGAP-USD4Log in for availability1 * 1 items - 66,20 €
Product Information
- Catalog #AvailabilityPrice
- Universal linear stand, for one single or multi-channel (8 / 12) pipetteUniversal linear stand, for one single or multi-channel (8 / 12) pipetteAXYGAP-USDLog in for availability1 * 1 items - 44,10 €



