ContactUsLinkComponent
KIT, GLOBAL PAN-METHYL HISTONE H3-K4 QUANTIFICATION KIT (FLUOROMETRIC), GLOBAL PAN-METHYL HISTONE H3-K4 QUANTIFICATION KIT (FLUOROME TRIC) IS USE FOR MEASURING GLOBAL MONO-, DI- AND TRI-METHYLATION OF HISTONE H3-K4. 1 * 1 KIT
Catalog # ABNOKA0591
Supplier:  Abnova
KIT, GLOBAL PAN-METHYL HISTONE H3-K4 QUANTIFICATION KIT (FLUOROMETRIC), GLOBAL PAN-METHYL HISTONE H3-K4 QUANTIFICATION KIT (FLUOROME TRIC) IS USE FOR MEASURING GLOBAL MONO-, DI- AND TRI-METHYLATION OF HISTONE H3-K4. 1 * 1 KIT
Catalog # ABNOKA0591
Supplier:  Abnova
Related Information