Combination pH electrodes, sensION™+
5014T, with PT1000 sensor, 0 - 14, −10...+100 °C, Glass, 3,0M KCl (refillable), 2 × Ceramic pin, 12×130 mm, BNC + banana, 1 m cable, General aqueous applications, TRIS buffers
Catalog # HACHLZW5014T97002
Supplier: Hach
Jump to:
Product Details & Documents
3-in-1 laboratory combination pH electrodes with built-in temperature sensor.
- 5010T and 5011T are low maintenance electrodes for general applications
- 5014T and 5021T are high performance electrodes for special or difficult applications
- For use with Hach sensION+ laboratory pH meters
Delivery information: Supplied with basic user manual. 5014T and 5021T also include 3,0 M KCl electrolyte refill solution (30 ml).
Specifications
- Connection:BNC + banana, 1 m cable
- Ø×L:12×130 mm
- For:General aqueous applications, TRIS buffers
- Type:5014T, with PT1000 sensor
- Material:Glass
- Seq:04
- Diaphr.:2 × Ceramic pin
- Electrolyte:3,0M KCl (refillable)
- Temp.:−10...+100 °C
- pH:0 - 14



