Order Entry
Export
Orders LinkContactUsLinkComponent
Safety helmets, Concept

Safety helmets, Concept

Safety helmet, vented, without ratchet, EU, Yellow
Catalog # 111-0869
Safety helmets, Concept
Catalog # 111-0869
Safety helmets, Concept
Safety helmets, Concept

Safety helmet, vented, without ratchet, EU, Yellow

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

Lightweight, comfortable and modern protective helmet made from ABS (acrylonitrile butadiene styrene) with large forehead area featuring an imprint for the company logo.

  • With extended nape area
  • Six point terylene inner cradle
  • Highly absorbent hydro-flock sweatband
  • With increased side rigidity (additional lateral deformity test)
  • Wide ventilation slots
  • Without ratchet, with full peak

Size: 510 to 630 mm

Weight: 325 g

EN 397

Certifications: EN 397

Documents

Specifications
  • Colour:
    Yellow
  • Description:
    Safety helmet, vented, without ratchet, EU
  • Seq:
    06