Order Entry
Export
Orders LinkContactUsLinkComponent
 

Buffer, TBS solution 10X concentrate (TRIS buffered saline) pH 7,4

TBS buffer solution 10X concentrate pH 7,4, 1 L
Catalog # CAYM600232-1
Supplier:   Cayman Chemical
Buffer, TBS solution 10X concentrate (TRIS buffered saline) pH 7,4
Catalog # CAYM600232-1
Supplier:  Cayman Chemical
CAS Number:  
Buffer, TBS solution 10X concentrate (TRIS buffered saline) pH 7,4

TBS buffer solution 10X concentrate pH 7,4, 1 L

CAS Number:  
Price
225,00 €
Log in for availability
1 * 1 L
Product Details & Documents

Formulation: Concentrated TBS (250 mM Tris-base; pH 7,4; 1,37 M sodium chloride and 27 mM KCl).

Suggested storage (long term) temperature: 4 °C.

Specifications

Specifications

  • Pk:
    1 L
  • Description:
    TBS buffer solution 10X concentrate pH 7,4

Specifications