Order Entry
Export
Orders LinkContactUsLinkComponent
Buffer, TBS solution 10X concentrate (TRIS buffered saline) pH 7,4

Buffer, TBS solution 10X concentrate (TRIS buffered saline) pH 7,4

TBS buffer solution 10X concentrate pH 7,4, 250 mL
Catalog # CAYM600232-250
Supplier:   Cayman Chemical
Buffer, TBS solution 10X concentrate (TRIS buffered saline) pH 7,4
Catalog # CAYM600232-250
Supplier:  Cayman Chemical
CAS Number:  
Buffer, TBS solution 10X concentrate (TRIS buffered saline) pH 7,4

TBS buffer solution 10X concentrate pH 7,4, 250 mL

CAS Number:  
Price
64,00 €
Log in for availability
1 * 250 mL
Product Details & Documents

Formulation: Concentrated TBS (250 mM Tris-base; pH 7,4; 1,37 M sodium chloride and 27 mM KCl).

Suggested storage (long term) temperature: 4 °C.

Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    TBS buffer solution 10X concentrate pH 7,4

Specifications