Buffers, potassium phosphate buffer (0,5 mol/L) solutions
Potassium phosphate buffer solution (0,5 mol/l), 8,0, Glass bottle, 1 L
Catalog # CAYM600209-1
Supplier: Cayman Chemical
Buffers, potassium phosphate buffer (0,5 mol/L) solutions
Catalog # CAYM600209-1
Supplier: Cayman Chemical
CAS Number:
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Description:Potassium phosphate buffer solution (0,5 mol/l)
- pH value:8,0



