Order Entry
Export
Orders LinkContactUsLinkComponent
Buffers, potassium phosphate buffer (0,5 mol/L) solutions

Buffers, potassium phosphate buffer (0,5 mol/L) solutions

Potassium phosphate buffer solution (0,5 mol/l), 7,0, Glass bottle, 250 ml
Catalog # CAYM600208-250
Supplier:   Cayman Chemical
Buffers, potassium phosphate buffer (0,5 mol/L) solutions
Catalog # CAYM600208-250
Supplier:  Cayman Chemical
CAS Number:  
Buffers, potassium phosphate buffer (0,5 mol/L) solutions

Potassium phosphate buffer solution (0,5 mol/l), 7,0, Glass bottle, 250 ml

CAS Number:  
Price
64,00 €
Log in for availability
1 * 250 mL
Product Details & Documents
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 ml
  • Description:
    Potassium phosphate buffer solution (0,5 mol/l)
  • pH value:
    7,0

Specifications