Sodium acetate
Glass bottle for solids, 0,82 g
Catalog # CAYM400037-100
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
Synonyms: Acetic acid sodium salt
- CAS Number:127-09-3
- Formula:H₃CCOONa
- MDL Number:MFCD00012459
- Molecular Weight:82.03 g/mol
- Merck Index:12,08711
- Boiling Pt:881.4 °C (1013 hPa)
- Melting Pt:324 °C
- Density:1.528 g/cm³ (20 °C)
- Flash Pt:>250 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:204-823-8
- REACH:01-2119485123-42
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:0,82 g



