Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
(±)-17-HDoHE
(±)-17-HDoHE
(±)-17-HDoHE
Print
(±)-17-HDoHE
250 µG
Catalog # CAYM33650-250
Supplier:
Cayman Chemical
(±)-17-HDoHE
Catalog # CAYM33650-250
Supplier:
Cayman Chemical
CAS Number:
90780-52-2
Expand image
Return to Top
(±)-17-HDoHE
250 µG
Specifications
Add to cart
Jump to:
Specifications
CAS Number:
90780-52-2
Price
792,00 €
Log in
for availability
Unit of Measure
1 * 250 µG
-
+
Add to cart
Other Product Family Options Available
Return Policy
Product Details & Documents
CAS Number:
90780-52-2
Formula:
C₂₂H₃₂O₃
Specifications
Specifications
Pk:
250 µG
Specifications