Order Entry
Export
Orders LinkContactUsLinkComponent
(±)-17-HDoHE

(±)-17-HDoHE

25 µG
Catalog # CAYM33650-25
Supplier:   Cayman Chemical
(±)-17-HDoHE
Catalog # CAYM33650-25
Supplier:  Cayman Chemical
CAS Number:  90780-52-2
(±)-17-HDoHE

25 µG

CAS Number:  90780-52-2
Price
106,00 €
Log in for availability
1 * 25 µG
Product Details & Documents
  • CAS Number:90780-52-2
  • Formula:C₂₂H₃₂O₃
Specifications

Specifications

  • Pk:
    25 µG

Specifications