Order Entry
Export
Orders LinkContactUsLinkComponent
2-Aminopyrazine

2-Aminopyrazine

Glass bottle for solids, 10 g
Catalog # CAYM10007093-10
Supplier:   Cayman Chemical
2-Aminopyrazine
Catalog # CAYM10007093-10
Supplier:  Cayman Chemical
CAS Number:  5049-61-6
2-Aminopyrazine

Glass bottle for solids, 10 g

CAS Number:  5049-61-6
Price
49,00 €
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  Aminopyrazine, Pyrazin-2-amine
  • CAS Number:5049-61-6
  • Formula:C₄H₅N₃
  • MDL Number:MFCD00006137
  • Molecular Weight:95.1 g/mol
  • Boiling Pt:203 °C (1013 hPa)
  • Melting Pt:117…120 °C
  • EINECS:225-748-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications