2-Aminopyrazine
Glass bottle for solids, 5 g
Catalog # CAYM10007093-5
Supplier: Cayman Chemical
Jump to:
Product Details & Documents
Synonyms: Aminopyrazine, Pyrazin-2-amine
- CAS Number:5049-61-6
- Formula:C₄H₅N₃
- MDL Number:MFCD00006137
- Molecular Weight:95.1 g/mol
- Boiling Pt:203 °C (1013 hPa)
- Melting Pt:117…120 °C
- EINECS:225-748-7
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



