Order Entry
Export
Orders LinkContactUsLinkComponent
 

Capsaicin

Glass bottle for solids, 1 g
Catalog # CAYM10010743-1
Supplier:   Cayman Chemical
Capsaicin
Catalog # CAYM10010743-1
Supplier:  Cayman Chemical
CAS Number:  404-86-4
Capsaicin

Glass bottle for solids, 1 g

CAS Number:  404-86-4
Price
87,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  8-Methyl-N-vanillyl-6-none-amid
  • CAS Number:404-86-4
  • Formula:C₁₈H₂₇NO₃
  • MDL Number:MFCD00017259
  • Molecular Weight:305.42 g/mol
  • Merck Index:12,01811
  • Boiling Pt:210…220 °C (0.01 mmHg)
  • Melting Pt:66.1 °C
  • EINECS:206-969-8
  • UN:2811
  • ADR:6.1,III

 

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications