Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Standards
Redox/ORP Standards
Redox standards
Redox standards
Print
Redox standards
300 mV standard, 500 mL
Catalog # JENW90238-50
Supplier:
Cole-Parmer
Redox standards
Catalog # JENW90238-50
Supplier:
Cole-Parmer
CAS Number:
Expand image
Return to Top
Redox standards
300 mV standard, 500 mL
Specifications
Add to cart
Jump to:
Specifications
CAS Number:
Price
610,00 €
Log in
for availability
Unit of Measure
1 * 500 mL
-
+
Add to cart
Other Product Family Options Available
Return Policy
Product Details & Documents
Specifications
Specifications
Pk:
500 mL
Description:
300 mV standard
Specifications