Order Entry
Export
Orders LinkContactUsLinkComponent
 

Needles, Spinocan®

Needles, Spinocan®, 18, 88 mm, 1,30 mm, Pink
  BRAU4501390-01
 :   B. Braun
Needles, Spinocan®
  BRAU4501390-01
 :  B. Braun
Needles, Spinocan®
Needles, Spinocan®

Needles, Spinocan®, 18, 88 mm, 1,30 mm, Pink

 
139,00 €
   
1 * 25 items
 
 
  • Colour code:
    Pink
  • Ø:
    1,30 mm
  • Gauge:
    18
  • Description:
    Needles, Spinocan®
  • Length:
    88 mm
  • Seq:
    002