Order Entry
Export
Orders LinkContactUsLinkComponent
 

Needles, Spinocan®

Needles, Spinocan®, 25, 88 mm, 0,50 mm, Orange
  BRAU4505905-01
 :   B. Braun
Needles, Spinocan®
  BRAU4505905-01
 :  B. Braun
Needles, Spinocan®
Needles, Spinocan®

Needles, Spinocan®, 25, 88 mm, 0,50 mm, Orange

 
139,00 €
   
1 * 25 items
 
 
  • Colour code:
    Orange
  • Ø:
    0,50 mm
  • Gauge:
    25
  • Description:
    Needles, Spinocan®
  • Length:
    88 mm
  • Seq:
    010